Learn More
Ace Glass, Inc. 14/10 75MM DIST COLUMN 14/10 75MM DIST COLUMN
SDP
Catalog No. 50976751
Encompass_Preferred
14/10 75mm long Vigreux distillation column. Length in millimeters refers to the effective length as measured from the lowest to the highest indent of the column.
Catalog No. 50-976-751 Supplier Ace Glass, Inc. Supplier No. 934532
14/10 75mm long Vigreux distillation column. Length in millimeters refers to the effective length as measured from the lowest to the highest indent of the column.