Learn More
Cayman Chemical 14 15-DIHET 100UG
SDP
Catalog No. 502896345
Encompass_Preferred
A urinary metabolite of (±)14(15)-EET which is elevated in pregnancy-induced hypertension
Catalog No. 50-289-6345 Supplier Cayman Chemical Supplier No. 51651100 UG
A urinary metabolite of (±)14(15)-EET which is elevated in pregnancy-induced hypertension