Learn More
Medchemexpress LLC (±)14(15)-DiHET-d11 | 98050-20-5 | 95.0% | C20H23D11O4 | 25 UG
SDP
Catalog No. 5000356761
Encompass_Preferred
(±)14,15-DiHETrE-d11 is the deuterium labeled metabolite of (±)14(15)-EET, formed by epoxide hydrolases. This compound is for research use only and not sold to patients. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer or internal standard
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-56761 Supplier Medchemexpress LLC Supplier No. 25UGHY143664S
(±)14,15-DiHETrE-d11 is the deuterium labeled metabolite of (±)14(15)-EET, formed by epoxide hydrolases. This compound is for research use only and not sold to patients. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer or internal standard
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS