Learn More
Cayman Chemical 14 15-EET methyl ester 100UG
SDP
Catalog No. 502892548
Encompass_Preferred
An esterified version of (±)14(15)-EET; more stable for long-term storage and can be readily hydrolyzed to the free acid just before use; equipotent to (±)14(15)-EET in inducing relaxation of isolated bovine coronary arteries precontracted by U-46619
Catalog No. 50-289-2548 Supplier Cayman Chemical Supplier No. 50650100 UG
An esterified version of (±)14(15)-EET; more stable for long-term storage and can be readily hydrolyzed to the free acid just before use; equipotent to (±)14(15)-EET in inducing relaxation of isolated bovine coronary arteries precontracted by U-46619