Learn More
Enzo Life Sciences 14,15-Leukotriene A4 methyl ester (50µg)
SDP
Supplier:  Enzo Life Sciences BMLLPA0040050
Encompass_Preferred
Biosynthesis in rabbit reticulocytes and human leukocytes. Alternative name: 14,15-LTA4-ME. Formulation: Oil dissolved in 2% triethylamine/hexane. Purity: ≥98% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-1163