Learn More
Enzo Life Sciences 14,15-Leukotriene C4 (50µg). CAS: 75290-60-7
SDP
Supplier:  Enzo Life Sciences BMLLPC0040050
Encompass_Preferred
Biosynthesis in RBL cells. Displays weak contractile activity on guinea pig parenchymal strip and ileum. Alternative name: 14,15-LTC4. Formulation: Oil dissolved in methanol/ammonium acetate buffer, 70/30, pH 5.6. Purity: ≥98% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-1165