Learn More
Enzo Life Sciences 14,15-Leukotriene D4 (50µg)
SDP
Supplier:  Enzo Life Sciences BMLLPD0040050
Encompass_Preferred
Novel leukotriene produced via the 15-LO pathway. Alternative name: 14,15-LTD4. Formulation: Oil dissolved in methanol/ammonium acetate buffer, 70/30, pH 5.6. Purity: ≥98% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-1166