Learn More
Cayman Chemical 14 15dehydro LeukotrIen B4
SDP
Catalog No. 502985024
Encompass_Preferred
A LTB4 receptor antagonist that has a higher binding affinity for BLT1 (Ki = 27 nM) compared to BLT2 (Ki = 473 nM); inhibits LTB4-induced release of lysozymes from rat polymorphonuclear leukoctyes with an IC50 value of 1 µM
Catalog No. 50-298-5024 Supplier Cayman Chemical Supplier No. 20150100 UG
A LTB4 receptor antagonist that has a higher binding affinity for BLT1 (Ki = 27 nM) compared to BLT2 (Ki = 473 nM); inhibits LTB4-induced release of lysozymes from rat polymorphonuclear leukoctyes with an IC50 value of 1 µM