Learn More
Cayman Chemical 14 15DiHETd11 GC MS Standards
SDP
Supplier:  Cayman Chemical 10008040100 UG
Encompass_Preferred
An internal standard for the quantification of 14(15)-DiHET by GC- or LC-MS
Catalog No. 50-288-7701