Learn More
Cayman Chemical 14 15DiHETd11 GC MS Standards
SDP
Catalog No. 502887700
Encompass_Preferred
An internal standard for the quantification of 14(15)-DiHET by GC- or LC-MS
Catalog No. 50-288-7700 Supplier Cayman Chemical Supplier No. 1000804050 UG
An internal standard for the quantification of 14(15)-DiHET by GC- or LC-MS