Learn More
Cayman Chemical 14 15EETd11 GC MS Standards
SDP
Catalog No. 502891043
Encompass_Preferred
An internal standard for the quantification of (±)14(15)-EET by GC- or LC-MS
Catalog No. 50-289-1043 Supplier Cayman Chemical Supplier No. 1000641025 UG
An internal standard for the quantification of (±)14(15)-EET by GC- or LC-MS