Learn More
Cayman Chemical 14 15EETd11 GC MS Standards
SDP
Supplier:  Cayman Chemical 1000641050 UG
Encompass_Preferred
An internal standard for the quantification of (±)14(15)-EET by GC- or LC-MS
Catalog No. 50-289-1044