Learn More
Cayman Chemical 14 15EETSI Fatty Acids and Der
SDP
Catalog No. 502888947
Encompass_Preferred
A metabolically stable methyl sulfonamide analog of 14(15)-EET; equipotent to 14(15)-EET in vascular agonist activity
Catalog No. 50-288-8947 Supplier Cayman Chemical Supplier No. 10009286100 UG
A metabolically stable methyl sulfonamide analog of 14(15)-EET; equipotent to 14(15)-EET in vascular agonist activity