Learn More
Cayman Chemical 14 15EpEDE Fatty Acids and Der
SDP
Catalog No. 502887987
Encompass_Preferred
An EpEDE acid formed from DGLA, either by a strong oxidizing agent or by cytochrome P450 epoxygenases; biological properties are poorly understood
Catalog No. 50-288-7987 Supplier Cayman Chemical Supplier No. 10007527500 UG
An EpEDE acid formed from DGLA, either by a strong oxidizing agent or by cytochrome P450 epoxygenases; biological properties are poorly understood