Learn More
Cayman Chemical 14 15Leukotriene D4
SDP
Catalog No. 502891076
Encompass_Preferred
A unique LT with weak contractile activity on both guinea pig ileum and pulmonary parenchyma compared to 5-LO-derived LTs; increases vascular permeability of human endothelial cell monolayers with similar potency to that of 5-LO-derived LTs
Catalog No. 50-289-1076 Supplier Cayman Chemical Supplier No. 10011361100 UG
A unique LT with weak contractile activity on both guinea pig ileum and pulmonary parenchyma compared to 5-LO-derived LTs; increases vascular permeability of human endothelial cell monolayers with similar potency to that of 5-LO-derived LTs