Learn More
Cayman Chemical 14 15Leukotriene D4
SDP
Supplier:  Cayman Chemical 1001136125 UG
Encompass_Preferred
A unique LT with weak contractile activity on both guinea pig ileum and pulmonary parenchyma compared to 5-LO-derived LTs; increases vascular permeability of human endothelial cell monolayers with similar potency to that of 5-LO-derived LTs
Catalog No. 50-289-1318