Learn More
MSC NEOPRN SPPRT CHEM RES GLV
Supplier:  MSC 74403825
Encompass
Chemical Resistant Gloves; Glove Type: General Purpose Chemical-Resistant ; Material: Neoprene ; Size: X-Large ; Numeric Size: 10 ; Coating Material: Neoprene ; Lining Material: Nylon
Catalog No. 50-273-7301