Learn More
Ace Glass, Inc. 14/20 100mm distillation column
SDP
Catalog No. 50976747
Encompass_Preferred
14/20 100mm long Vigreux distillation column. Length in millimeters refers to the effective length as measured from the lowest to the highest indent of the column.
Catalog No. 50-976-747 Supplier Ace Glass, Inc. Supplier No. 934508
14/20 100mm long Vigreux distillation column. Length in millimeters refers to the effective length as measured from the lowest to the highest indent of the column.