Learn More
Ace Glass, Inc. Thermometer adapter, PTFE, 14/20 to compression seal, use with 6-7mm diameter tubes, Viton; O-rings
SDP
Catalog No. 509558457
Encompass_Preferred
14/20 PTFE thermometer adapter with FETFE o-ring, for use with plain stem thermometers, gas inlet tubes, etc. The o-ring compression seal on the adapter allows adjustable depth positioning. Adapter accomm ODates tubes up to 7mm.
Catalog No. 50-955-8457 Supplier Ace Glass, Inc. Supplier No. 830007
14/20 PTFE thermometer adapter with FETFE o-ring, for use with plain stem thermometers, gas inlet tubes, etc. The o-ring compression seal on the adapter allows adjustable depth positioning. Adapter accomm ODates tubes up to 7mm.