Learn More
Chemglass Life Sciences Adapter, Connecting, Airfree, Schlenk, (1) 14/20 Outer Joint, (2) 14/20 Inner Joints
SDP
Catalog No. AF050201
Encompass_Preferred
Adapter, Connecting, Airfree, Schlenk, (1) 14/20 Outer Joint, (2) 14/20 Inner Joints. A 3-way adapter for the connection of 3 vessels. With a top outer and two lower inner joints, all being the same size.
Catalog No. AF050201 Supplier Chemglass Life Sciences Supplier No. AF050201
Adapter, Connecting, Airfree, Schlenk, (1) 14/20 Outer Joint, (2) 14/20 Inner Joints. A 3-way adapter for the connection of 3 vessels. With a top outer and two lower inner joints, all being the same size.