Learn More
Chemglass Life Sciences Adapter, Flushing, 14/20 Joint Size, 4mm Glass Stpk, Airfree, Schlenk
SDP
Supplier:  Chemglass Life Sciences AF0509A01
Encompass_Preferred
Adapter, Flushing, 14/20 Joint Size, 4mm Glass Stpk, Airfree, Schlenk. Similar to AF-0509 but having a top outer joint and lower inner joint of the same size. Addition of the outer joint permits use of the larger sizes of the CG-3022 septum stoppers for repeated needle penetrations. Adapter also allows for evacuation and transfer of product into another vessel with the same size standard taper joints. Available with a choice of a 4mm glass high vacuum stopcock having a Stainless Steel constant pressure retention device or 4mm PTFE stopcock. PLEASE NOTE: NOT SUPPLIED WITH SEPTUM STOPPER.
Catalog No. AF0509A01