Learn More
Ace Glass, Inc. Adapter, stopcock, angled hose connection, PTFE plug, 14/20
SDP
Catalog No. 509558648
Encompass_Preferred
14/20 stopcock adapter with an angled hose connection and a PTFE plug.
Catalog No. 50-955-8648 Supplier Ace Glass, Inc. Supplier No. 908012
14/20 stopcock adapter with an angled hose connection and a PTFE plug.