Learn More
Caplugs 14/20 CAP PLUG MINI 1000PK
Catalog No. NC9032050
Encompass_Preferred
Tapered Plug, With Wide Thick Flange, Color: Yellow, Material: Low Density Polyethylene, Specifications: NAS-816, No of Pieces: 5000, 1000/PK, Shipping Weight: 11.690 kg
This item is not returnable. View return policy
Catalog No. NC9032050 Supplier Caplugs Supplier No. 99191374
Tapered Plug, With Wide Thick Flange, Color: Yellow, Material: Low Density Polyethylene, Specifications: NAS-816, No of Pieces: 5000, 1000/PK, Shipping Weight: 11.690 kg