Learn More
Ace Glass, Inc. Joint clips, 14/20, Keck; type, POM acetal resin, not for positive pressure use, yellow, pk/10
SDP
Catalog No. 509557890
Encompass
14/20 yellow Keck type joint clips made from POM acetal resin. Clips will not scratch the glass, and are resistant to concentrated alkalies and dilute acids. Useful temp range: -40 to 140 C. Not for pressure applications. Pk/10.
Catalog No. 50-955-7890 Supplier Ace Glass, Inc. Supplier No. 759814
14/20 yellow Keck type joint clips made from POM acetal resin. Clips will not scratch the glass, and are resistant to concentrated alkalies and dilute acids. Useful temp range: -40 to 140 C. Not for pressure applications. Pk/10.