Learn More
Ace Glass, Inc. Distillation head, automatic reflux control via solenoid valve arrangement.
SDP
Catalog No. 50979991
Encompass
Distillation head featuring an automatic reflux control by means of a solenoid valve arrangement. Supplied with a #7 Ace-Thred™ bushing for use with 6-7mm diameter temperature probes or thermometers. Equalizing arm 12/3 joints are supplied with ACE 7669-03 clamps. Top and bottom joints are 14/20. *Thermometer not included.
Catalog No. 50-979-991 Supplier Ace Glass, Inc. Supplier No. 936208
Distillation head featuring an automatic reflux control by means of a solenoid valve arrangement. Supplied with a #7 Ace-Thred™ bushing for use with 6-7mm diameter temperature probes or thermometers. Equalizing arm 12/3 joints are supplied with ACE 7669-03 clamps. Top and bottom joints are 14/20. *Thermometer not included.