Learn More
Chemglass Life Sciences 14/20 Inner Joint, Un-Cut Drip, on 12.7mm OD x 1.6mm
SDP
Catalog No. CG11701
Encompass_Preferred
Full or medium length standard taper inner joint having a drip tip. Joints are available uncut or with the drip tip being cut at a 45º angle. Tooled on the same tubing sizes as the CG-100 inner joints. 14/20 Inner Joint, Un-Cut Drip, on 12.7mm OD x 1.6mm.
Catalog No. CG11701 Supplier Chemglass Life Sciences Supplier No. CG11701
Full or medium length standard taper inner joint having a drip tip. Joints are available uncut or with the drip tip being cut at a 45º angle. Tooled on the same tubing sizes as the CG-100 inner joints. 14/20 Inner Joint, Un-Cut Drip, on 12.7mm OD x 1.6mm.