Learn More
Chemglass Life Sciences 14/20 Inner Joint, Medium Length, on 13mm OD x 2.2mm W tubing
SDP
Catalog No. CG10408
Encompass_Preferred
Adapter, Thermometer, Inlet, 10° Offset, 29/26 Inner Joint. Adapter has a #7 Chem-Thread at the top for vacuum tight seal of plain stem thermometers or any other tube having an O.D. between 4 and 7mm. When used with vertical side neck flasks, thermometers or bleed tubes can be angled into the bottom of the flask. Supplied complete with a compression cap and Viton o-ring.
Catalog No. CG10408 Supplier Chemglass Life Sciences Supplier No. CG10408
Adapter, Thermometer, Inlet, 10° Offset, 29/26 Inner Joint. Adapter has a #7 Chem-Thread at the top for vacuum tight seal of plain stem thermometers or any other tube having an O.D. between 4 and 7mm. When used with vertical side neck flasks, thermometers or bleed tubes can be angled into the bottom of the flask. Supplied complete with a compression cap and Viton o-ring.