Learn More
Chemglass Life Sciences 14/20 Outer Joint, Dewar Seal, 35mm OD Flare
SDP
Catalog No. CG13132
Encompass_Preferred
14/20 Outer Joint, Dewar Seal, 35mm OD Flare. Used in construction of apparatus requiring the outer member to be jacketed. Configuration of flare permits seal to be made without having to regrind the joint.
Catalog No. CG13132 Supplier Chemglass Life Sciences Supplier No. CG13132
14/20 Outer Joint, Dewar Seal, 35mm OD Flare. Used in construction of apparatus requiring the outer member to be jacketed. Configuration of flare permits seal to be made without having to regrind the joint.