Learn More
Ace Glass, Inc. Adapter, #7 Ace-Thred top, 14/20 inner bottom, nylon bushing with FETFE O-ring
SDP
Supplier:  Ace Glass, Inc. 502826
Encompass_Preferred
#7 Ace-Thred mini adapter with 14/20 inner joint bottom, and nylon bushing with FETFE O-ring. Used for probes, thermometers, bleed tubes, and electrodes. Adapter will accomm ODate items up to 7mm diameter. Suitable for vacuum work.
Catalog No. 50-955-6899