Learn More
Chemglass Life Sciences 14/20 Outer Rodavissr Joint
SDP
Catalog No. CG17201
Encompass_Preferred
14/20 Outer Rodaviss Joint. Rodaviss Outer Joint is threaded to accept CG-182 caps.
Catalog No. CG17201 Supplier Chemglass Life Sciences Supplier No. CG17201
14/20 Outer Rodaviss Joint. Rodaviss Outer Joint is threaded to accept CG-182 caps.