Learn More
Ace Glass, Inc. 14/20 PTFE SLEEVE 3/PK 14/20 PTFE SLEEVE 3/PK
SDP
Catalog No. 509557920
Encompass
PTFE sleeves with 14/20 joint size and .4mm wall thickness. Sturdy and reusable, features reinforced gripping ring for easy and safe removal from standard taper ground joints. Pack of 3.
Catalog No. 50-955-7920 Supplier Ace Glass, Inc. Supplier No. 764104
PTFE sleeves with 14/20 joint size and .4mm wall thickness. Sturdy and reusable, features reinforced gripping ring for easy and safe removal from standard taper ground joints. Pack of 3.