Learn More
Ace Glass, Inc. 14/20 PTFE SLEEVES/PK 12 14/20 PTFE SLEEVES/PK 12
SDP
Catalog No. NC9619557
Encompass
14/20 PTFE standard taper joint sleeves. Sleeves are inserted between standard taper ground glass inner and outer joints to help prevent sticking or the need for lubrication. Thickness .050mm, Max temp 280 deg.C.
Catalog No. NC9619557 Supplier Ace Glass, Inc. Supplier No. 7643109
14/20 PTFE standard taper joint sleeves. Sleeves are inserted between standard taper ground glass inner and outer joints to help prevent sticking or the need for lubrication. Thickness .050mm, Max temp 280 deg.C.