Learn More
Chemglass Life Sciences 14/20 Side Seal Joints, Sidearm OD: 6.35mm, Hole Size: 3mm
SDP
Catalog No. CG13201
Encompass_Preferred
14/20 Side Seal Joints, Sidearm OD: 6.35mm, Hole Size: 3mm. Outer joint having a tubulation sealed at the midpoint of the ground portion, and a corresponding hole drilled in the inner joint permitting venting of the side tube when the inner joint is turned to the proper position.
Catalog No. CG13201 Supplier Chemglass Life Sciences Supplier No. CG13201
14/20 Side Seal Joints, Sidearm OD: 6.35mm, Hole Size: 3mm. Outer joint having a tubulation sealed at the midpoint of the ground portion, and a corresponding hole drilled in the inner joint permitting venting of the side tube when the inner joint is turned to the proper position.