Learn More
Chemglass Life Sciences Septum Stopper, Sleeve Type, For 14/20-14/35 Outer Joints, White
SDP
Supplier:  Chemglass Life Sciences CG302297
Encompass_Preferred
Septum Stopper, Sleeve Type, For 14/20-14/35 Outer Joints, White. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.
Catalog No. CG302297