Learn More
MSC Baldwin Air Filter Element, 6 in OD, 1 ea
Catalog No. 5000503856
Encompass
Baldwin axial seal air filter elements are engineered to protect equipment by effectively preventing airborne contaminants from entering the combustion chamber. This ensures proper engine function and extends the life of critical components by maintaining a clean air supply.
  • Outer air element with lift tabs
  • Compatible with PA2306 systems
  • Suitable for Allis Chalmers and Ford equipment
  • Features an attached gasket
Catalog No. 50-005-03856 Supplier MSC Supplier No. 10942217
Baldwin axial seal air filter elements are engineered to protect equipment by effectively preventing airborne contaminants from entering the combustion chamber. This ensures proper engine function and extends the life of critical components by maintaining a clean air supply.
  • Outer air element with lift tabs
  • Compatible with PA2306 systems
  • Suitable for Allis Chalmers and Ford equipment
  • Features an attached gasket