Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 14-3-3 epsilon antibody (YA2629) | 10 UL
SDP

Catalog No. 5000443542
Encompass_Preferred

This 14-3-3 epsilon antibody (YA2629) is a rabbit-derived, non-conjugated IgG monoclonal antibody that specifically targets the 14-3-3 epsilon protein. It is suitable for a diverse array of immunological applications.

  • Host species: rabbit
  • Reactivity: human, mouse, rat
  • Conjugation: non-conjugated
  • Predicted and observed molecular weight: 29 kDa
  • Purity: affinity chromatography
  • Storage: -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Appearance: liquid
  • Applications include Western Blot (WB), Immunohistochemistry (IHC-P), Immunocytochemistry/Immunofluorescence (ICC/IF), and Flow Cytometry (FC)
  • Formulation: rabbit IgG in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • SwissProt ID: P62258
  • Research field: neuroscience

Catalog No. 50-004-43542 Supplier Medchemexpress LLC Supplier No. HYP8288410UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This 14-3-3 epsilon antibody (YA2629) is a rabbit-derived, non-conjugated IgG monoclonal antibody that specifically targets the 14-3-3 epsilon protein. It is suitable for a diverse array of immunological applications.

  • Host species: rabbit
  • Reactivity: human, mouse, rat
  • Conjugation: non-conjugated
  • Predicted and observed molecular weight: 29 kDa
  • Purity: affinity chromatography
  • Storage: -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Appearance: liquid
  • Applications include Western Blot (WB), Immunohistochemistry (IHC-P), Immunocytochemistry/Immunofluorescence (ICC/IF), and Flow Cytometry (FC)
  • Formulation: rabbit IgG in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • SwissProt ID: P62258
  • Research field: neuroscience

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.