Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 14-3-3 epsilon Antibody (YA2629) | 50 UL
SDP

Catalog No. 5000443543
Encompass_Preferred

14-3-3 epsilon Antibody (YA2629) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to 14-3-3 epsilon. This protein serves as an adapter implicated in the regulation of a diverse array of both general and specialized signaling pathways, binding to numerous partners through the recognition of phosphoserine or phosphothreonine motifs, thereby modulating the activity of the binding partner.

  • Supplied in rabbit IgG in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA.
  • Appearance: Liquid
  • Applications & Dilution Ratios: WB: 1:500-1:1000, IHC-P: 1:50-1:100, ICC/IF: 1:50-1:200, FC: 1:50-1:100
  • Shipping with blue ice.

Catalog No. 50-004-43543 Supplier Medchemexpress LLC Supplier No. HYP8288450UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

14-3-3 epsilon Antibody (YA2629) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to 14-3-3 epsilon. This protein serves as an adapter implicated in the regulation of a diverse array of both general and specialized signaling pathways, binding to numerous partners through the recognition of phosphoserine or phosphothreonine motifs, thereby modulating the activity of the binding partner.

  • Supplied in rabbit IgG in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA.
  • Appearance: Liquid
  • Applications & Dilution Ratios: WB: 1:500-1:1000, IHC-P: 1:50-1:100, ICC/IF: 1:50-1:200, FC: 1:50-1:100
  • Shipping with blue ice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.