Learn More
Medchemexpress LLC 14-3-3 epsilon protein, mouse | 98.0% | 10 UG
SDP
Catalog No. 5000293241
Encompass_Preferred
Recombinant mouse 14-3-3 epsilon protein with an N-terminal 6xHis tag, expressed in Escherichia coli and supplied as a purified protein in a 10 μg vial. The protein has an expected molecular weight of ~31 kDa and is provided with QC documentation for research use in protein-protein interaction studies, antibody development, and assay setup.
  • Purity ≥98% as determined by reducing SDS-PAGE.
  • N-terminal 6xHis tag for affinity purification and detection.
  • Expressed in Escherichia coli to provide recombinant protein material.
  • Approximate molecular weight ~31 kDa for gel and mass confirmation.
  • Supplied as a low-quantity vial suitable for assay optimization and pilot studies.
  • Provided with COA/SDS documentation for traceability and handling.
Catalog No. 50-002-93241 Supplier Medchemexpress LLC Supplier No. HYP7554910UG
Recombinant mouse 14-3-3 epsilon protein with an N-terminal 6xHis tag, expressed in Escherichia coli and supplied as a purified protein in a 10 μg vial. The protein has an expected molecular weight of ~31 kDa and is provided with QC documentation for research use in protein-protein interaction studies, antibody development, and assay setup.
  • Purity ≥98% as determined by reducing SDS-PAGE.
  • N-terminal 6xHis tag for affinity purification and detection.
  • Expressed in Escherichia coli to provide recombinant protein material.
  • Approximate molecular weight ~31 kDa for gel and mass confirmation.
  • Supplied as a low-quantity vial suitable for assay optimization and pilot studies.
  • Provided with COA/SDS documentation for traceability and handling.