Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 14-3-3 epsilon protein, mouse (his) | >95.0% | 500 UG
SDP

Catalog No. 5000292984
Encompass_Preferred

Recombinant mouse 14-3-3 epsilon protein with an N-terminal 6xHis tag, expressed in Escherichia coli and supplied lyophilized. Purified to >95% and with an approximate molecular weight of 31 kDa, the product is formulated for research use; consult the SDS and COA for handling, reconstitution, and safety information.

  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Expressed in Escherichia coli and supplied lyophilized for stability.
  • Contains an N-terminal 6xHis tag for affinity purification or detection.
  • Molecular weight approximately 31 kDa.
  • Formulated in Tris-HCl (50 mM) and NaCl (300 mM), 0.22 μm filtered prior to lyophilization.
  • Reconstitute to at least 100 μg/mL, aliquot, and store at -20°C or -80°C for long-term storage.

Catalog No. 50-002-92984 Supplier Medchemexpress LLC Supplier No. HYP75549500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse 14-3-3 epsilon protein with an N-terminal 6xHis tag, expressed in Escherichia coli and supplied lyophilized. Purified to >95% and with an approximate molecular weight of 31 kDa, the product is formulated for research use; consult the SDS and COA for handling, reconstitution, and safety information.

  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Expressed in Escherichia coli and supplied lyophilized for stability.
  • Contains an N-terminal 6xHis tag for affinity purification or detection.
  • Molecular weight approximately 31 kDa.
  • Formulated in Tris-HCl (50 mM) and NaCl (300 mM), 0.22 μm filtered prior to lyophilization.
  • Reconstitute to at least 100 μg/mL, aliquot, and store at -20°C or -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.