Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 14-3-3 epsilon protein, mouse (His) | ≥98.0% | 50 UG
SDP

Catalog No. 5000292985
Encompass_Preferred

Recombinant mouse 14-3-3 epsilon His-tag protein supplied as a lyophilized powder for research use. The protein is produced in Escherichia coli, carries an N-terminal 6xHis affinity tag, and has an approximate molecular weight of 31 kDa. Supplied with high purity and low endotoxin for biochemical assays and binding studies.

  • Recombinant protein expressed in Escherichia coli.
  • N-terminal 6xHis tag for affinity purification and detection.
  • Lyophilized powder formulation for extended shelf life.
  • Approximate molecular weight 31 kDa.
  • Purity ≥98.0% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL), suitable for in vitro assays.
  • Formulated in Tris-HCl, NaCl; reconstitute at ≥100 μg/mL.
  • Store lyophilized at -20°C; freeze aliquots at -80°C for long term.

Catalog No. 50-002-92985 Supplier Medchemexpress LLC Supplier No. HYP7554950UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse 14-3-3 epsilon His-tag protein supplied as a lyophilized powder for research use. The protein is produced in Escherichia coli, carries an N-terminal 6xHis affinity tag, and has an approximate molecular weight of 31 kDa. Supplied with high purity and low endotoxin for biochemical assays and binding studies.

  • Recombinant protein expressed in Escherichia coli.
  • N-terminal 6xHis tag for affinity purification and detection.
  • Lyophilized powder formulation for extended shelf life.
  • Approximate molecular weight 31 kDa.
  • Purity ≥98.0% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL), suitable for in vitro assays.
  • Formulated in Tris-HCl, NaCl; reconstitute at ≥100 μg/mL.
  • Store lyophilized at -20°C; freeze aliquots at -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.