Learn More
Enzo Life Sciences 14-3-3 eta polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA4760100
Encompass_Preferred
Immunogen: Acetylated N-terminal domain of 14-3-3 eta. Species reactivity: Human, Mouse, Rat, Sheep, Chicken, Bovine. Applications: IHC, WB. Formulation: Liquid. Neat serum containing sodium azide.
Catalog No. 50-201-1833