Learn More
Cell Signaling Technology 14-3-3 Family Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502729648
Encompass
The 14-3-3 Family Antibody Sampler Kit provides an economical means to investigate the expression of various 14-3-3 isoforms within the cell. This kit contains a selection of primary antibodies against 14-3-3 isoforms and an anti-rabbit IgG secondary antibody, providing sufficient material to perform two Western blot experiments.
  • Designed for the investigation of 14-3-3 isoform expression
  • Includes primary antibodies for 14-3-3 beta/alpha, gamma, zeta/delta, epsilon, tau, and eta
  • Supplied with HRP-linked anti-rabbit IgG secondary antibody
  • Optimized for Western blotting applications
Catalog No. 50-272-9648 Supplier Cell Signaling Technology Supplier No. 9769T
The 14-3-3 Family Antibody Sampler Kit provides an economical means to investigate the expression of various 14-3-3 isoforms within the cell. This kit contains a selection of primary antibodies against 14-3-3 isoforms and an anti-rabbit IgG secondary antibody, providing sufficient material to perform two Western blot experiments.
  • Designed for the investigation of 14-3-3 isoform expression
  • Includes primary antibodies for 14-3-3 beta/alpha, gamma, zeta/delta, epsilon, tau, and eta
  • Supplied with HRP-linked anti-rabbit IgG secondary antibody
  • Optimized for Western blotting applications