Learn More
Enzo Life Sciences 14-3-3 polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA4830100
Encompass_Preferred
Immunogen: Recombinant human 14-3-3. Species reactivity: Yeast, Plant, Mammalian, Invertebrate, Chicken. Applications: IHC, IP, WB. Formulation: Liquid. Neat serum.
Catalog No. 50-201-1838