Learn More
Enzo Life Sciences 14-3-3 zeta (human), (recombinant) (GST-tag) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE3180100
Encompass_Preferred
Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS/HCl, pH 7.5, 150mM NaCl. Source: Produced in E. coli. Human 14-3-3ζ is fused at the N-terminus to a GST-tag. Long Term Storage: -80°C.
Catalog No. 50-201-1979