Learn More
Enzo Life Sciences 14-3-3 epsilon (human), (recombinant) (untagged) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2090100
Encompass_Preferred
Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 20 mM Tris-HCl, pH 8, containing 100 mM NaCl. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-3996