Learn More
Enzo Life Sciences 14-3-3zeta polyclonal antibody (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA4820025
Encompass_Preferred
Immunogen: Acetylated N-terminal epitope of 14-3-3 zeta. Species reactivity: Human, Mouse, Rat, Sheep, Chicken, Bovine. Applications: IHC, WB Affinity chromatography. Formulation: Liquid. Neat serum containing sodium azide.
Catalog No. 50-201-1836