Learn More
Chemglass Life Sciences 14/35 Outer Joint, 19mm X 1.8W Tubing
SDP
Catalog No. 501531889
Encompass_Preferred
14/35 Outer Joint, 19mm X 1.8W Tubing. Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint.
Catalog No. 50-153-1889 Supplier Chemglass Life Sciences Supplier No. CGB10104
14/35 Outer Joint, 19mm X 1.8W Tubing. Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint.