Learn More
Chemglass Life Sciences 14/35 Outer Joint, Full Length, on 19mm OD x 1.8mm W tubing
SDP
Catalog No. CG10104
Encompass_Preferred
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint. 14/35 Outer Joint, Full Length, on 19mm OD x 1.8mm W tubing.
Catalog No. CG10104 Supplier Chemglass Life Sciences Supplier No. CG10104
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint. 14/35 Outer Joint, Full Length, on 19mm OD x 1.8mm W tubing.