Learn More
Chemglass Life Sciences Replacement Nuts for 14.4/44 PTFE Plug
SDP
Catalog No. CG45712
Encompass_Preferred
Replacement Nuts for 14.4/44 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.
Catalog No. CG45712 Supplier Chemglass Life Sciences Supplier No. CG45712
Replacement Nuts for 14.4/44 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.