Learn More
Chemglass Life Sciences Replacement Washers for 14.4/44 PTFE Plug
SDP
Catalog No. CG45711
Encompass_Preferred
Replacement Washers for 14.4/44 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.
Catalog No. CG45711 Supplier Chemglass Life Sciences Supplier No. CG45711
Add to Cart
Add to Cart
Replacement Washers for 14.4/44 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.